Condenser, Friedrichs, 'Ace-Safe' Connectors, Ace Glass
Supplier: Ace Glass
5971-111 5971-116 5971-124 5971-128
89053-874EA 780.28 CAD
89053-874 89053-876 89053-878 89053-880
Condenser, Friedrichs, 'Ace-Safe' Connectors, Ace Glass
Condensers
This Friedrichs condenser has a molded spiral condensing surface, angled standard taper reinforced outer joint at top and vertical standard taper inner joint at bottom
Fabricated from borosilicate glass. Jacket length is 250mm. Has two Ace-Thred connections at top for use with 1/4-inch I.D. tubing.
Ordering information: Includes two #7 "Ace-Safe" Connectors, Order Code 89052-936.