Condenser, Friedrichs, Hose Barbs, Ace Glass Incorporated
Supplier: Ace Glass
5971-10 5971-15 5971-20 5971-23 5971-27
89053-872EA 699.73 CAD
89053-872 89053-882 89053-884 89053-886 89053-888
Condenser, Friedrichs, Hose Barbs, Ace Glass Incorporated
Condensers
This Friedrichs condenser has a molded spiral condensing surface, angled standard taper reinforced outer joint at top and vertical standard taper inner joint at bottom
Fabricated from borosilicate glass. Jacket length is 250mm. Has two hose barbs for use with 3/8-inch I.D. tubing, size D hose connections.