Cold Finger for Distillation Heads and Systems, Ace Glass Incorporated
Supplier: Ace Glass
9250-02 9250-04 9250-08
89074-304EA 228.3 CAD
89074-304 89074-306 89074-308
Cold Finger for Distillation Heads and Systems, Ace Glass Incorporated
Distillation Components Distillation Heads
Cold Finger accessory for standard distillation heads and systems
Fabricated from borosilicate glass. This cold finger has a standard taper inner joint that fits inside the upper outer joint and tube to provide added cooling ability and faster condensation. Has two upper, glass, size A hose connections that take 5/16-inch tubing.
Ordering information: Fits into 89055-700, 89055-694, and 89055-690 distilling heads.