Dewar Condenser, Ace Glass Incorporated
Supplier: Ace Glass
5964-13 5964-14 5964-12
89053-864EA 757.66 CAD
89053-864 89053-842 89053-930
Dewar Condenser, Ace Glass Incorporated
Condensers
Dewar type condenser for use with dry ice or other solid cooling agents
Upper tube is 12mm O.D. At bottom is standard taper 24/40 inner joint.