Suba·Seal® Septa, Ace Glass
Supplier: Ace Glass
9107-110 9107-111 9107-123 9107-124 9107-130 9107-131 9107-134 9107-135 9107-142 9107-143 9107-154 9107-155
89166-128PK 273.39 CAD
89166-128 89166-130 89166-132 89166-134 89166-136 89166-138 89166-140 89166-142 89166-144 89166-146 89166-148 89166-150
Suba·Seal® Septa, Ace Glass
Stoppers Rubber Stoppers
The Suba·Seal® Septa is a high quality rubber septa designed with serrated bottom for sealing open-top glass vessels as well as standard taper joints
Suba·Seal® Septa have serrated sealing capability to the inside of the vessel, as well as the turn-over top seal. Autoclaveable and packaged in white-room conditions.
Ideal for moisture or air sensitive applications.
Ordering information: Suba-Seal® is a trademark of Sigma-Aldrich Biotechnology LP.