PRP-1 Polymeric Reversed Phase HPLC Columns, Hamilton Company

Supplier: Hamilton
79444 79422 79425 79423 79427 79447 79445 79380 79366 79571 79426 79317 79351 79352 79381 79428 79443 79478 79496 79499 79531 79479 79495 79529
21480-015EA 1146.76 CAD
21480-015 21480-016 21480-017 21480-018 21480-050 21493-100 21493-225 21493-446 21493-464 89168-728 21480-120 89221-062 89221-068 89221-070 89221-098 89221-112 89221-116 89221-138 89221-146 89221-148 89221-152 89221-140 89221-144 10040-432
PRP-1 Polymeric Reversed Phase HPLC Columns, Hamilton Company
Chromatography Columns
Hamilton Company offers one of the most comprehensive selections of chromatography columns in the industry.

  • pH Stable from 1 to 13
  • Polymeric Columns for General Separations
  • Four Particle Sizes: 5, 7, 10, and 12 to 20 µm
  • Custom Hardware Dimensions Available
  • Analytical and Semiprep/Preparative Guard Columns
  • Buffer strength 0.0 - 0.5N

Each column is manufactured under rigorous criteria and tested for resolution and retention time accuracy.

PRP-1 polymeric HPLC columns with 100 Å pore size particles are designed for broad ranging uses, similar to C8 orMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR