Condenser, Reflux, Coiled, Ace Glass Incorporated
Supplier: Ace Glass
5955-13 5955-34 5955-15 5955-14
89053-790EA 1061.59 CAD
89053-790 89053-846 89053-902 89054-002
Condenser, Reflux, Coiled, Ace Glass Incorporated
Condensers
Double jacketed, coiled reflux condenser with standard taper inner joint at bottom and standard taper reinforced outer joint at top
fabricated from borosilicate glass. Use with 3/8-inch I.D. tubing, size D hose connections