Condenser, Reflux, Coiled, Hose Barbs, Ace Glass Incorporated
Supplier: Ace Glass
5953-12 5953-13 5953-14 5953-15
89053-832EA 608.64 CAD
89053-832 89053-834 89053-836 89053-838
Condenser, Reflux, Coiled, Hose Barbs, Ace Glass Incorporated
Condensers
Coiled reflux condensing tube, with standard taper 24/40 inner joint at bottom and standard taper 24/40 reinforced outer joint at top
Fabricated from borosilicate glass. Use with 3/8-inch I.D. tubing, size D hose connections.
Order Now
SPECIFICATIONS
Environmentally Preferable