Condenser, Allihn, with No. 7 Ace-Thred Connections
Supplier: Ace Glass
5945-76 5946-116 5946-16 5946-118 5946-18 5946-122 5946-22
89053-810EA 1328.9 CAD
89053-810 89053-812 89053-852 89053-920 89053-926 89214-960 89214-968
Condenser, Allihn, with No. 7 Ace-Thred Connections
Condensers
Bulb type Allihn condenser, approximately one bulb per 50 mm, with inner and outer joint at bottom and top
Fabricated from borosilicate glass. No.7 Ace-Thred hose connections on inlet and outlet with barb for 1/4-inch I.D. tubing.
Ordering information: Available with or without "Ace-Safe" Connectors.