Condenser, 'A' Assembly, for Rotary Evaporators, Ace Glass
Supplier: Ace Glass
3950-03 3950-01
89083-106EA 2212.55 CAD
89083-106 89083-110
Condenser, 'A' Assembly, for Rotary Evaporators, Ace Glass
Condensers
A Assembly angled condenser with inner coil for many popular bench-scale rotary evaporators
Fabricated from borosilicate glass. Fits models 200/205 and Series 114-144. Available poly coated for safety, or uncoated. Also available complete with 1000mL receiving flask and stainless steel clamp. The bottom joint is spherical [SJ] 35/20.
Ordering information: Includes o-ring, 89059-774 GL thread caps, and 89059-810 angled hose connectors. The bottom joint is spherical [SJ] 35/20.