Condenser, Allihn, Jacketed
Supplier: Ace Glass
5941-12 5941-14
89053-782EA 361.72 CAD
89053-782 89053-792
Condenser, Allihn, Jacketed
Condensers
Bulb type Allihn condenser, approximately one bulb per every 50 mm, with 24/40 standard taper inner joint at bottom, open tube at the top
Fabricated from borosilicate glass. Use with 3/8-inch I.D. tubing, size D hose connections.
Order Now
SPECIFICATIONS
Environmentally Preferable