FreeZone® Product Shelves, Labconco®
Supplier: Labconco
7317100 7317200
75840-998EA 1888.1 CAD
75840-998 75841-000
FreeZone® Product Shelves, Labconco®
Furniture Shelves
Product Shelves fit inside FreeZone® Clear or Port Drying Chambers to accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications.
  • Removable, 10" Ø aluminum shelves with ¼" rim slide into the stainless steel support
  • Product shelves are designed to fit inside drying chambers
  • Shelf can be placed in freezer to pre-freeze sample for quick loading
  • The removable shelf design allows for flexible shelf height spacing
Also available are heated shelves that include ambient to +40 °C (+104 °F) variable heat control on modelsMore Product Information