VWR® Sealing Film, Polyester
Supplier: VWR International
THNL100 THSNL100
89134-430CS 162.78 CAD
89134-430 89134-432
VWR® Sealing Film, Polyester
Seals Microplate Seals
Polyester film, acrylic adhesive. These 49.5 µm thick polyester films minimise evaporation and prevent spillage and contamination between wells.
  • Recommended temperature range from −40 to +120 °C
  • Available sterile and non sterile
  • Marginally pierceable by single channel pipette
  • Applications: Short-term storage, incubation, ELISA
Each film L×D: 141,0×79,4 mm