FreeZone® Heated Shelves with Sensors, Labconco®
Supplier: Labconco
7760800 7760801
14221-948EA 2133 CAD
14221-948 14221-950
FreeZone® Heated Shelves with Sensors, Labconco®
Furniture Shelves
Heated shelf with sensor, for use with bulk tray dryer.
  • 200-watt heater ensures efficient heating from –20 to 60 °C
A microprocessor in the bulk tray dryer maintains system temperature within 3 °C of set point.