Guardian™ 500 Airflow Monitor with Epoxy-Coated Steel Casing, Labconco®
Supplier: Labconco
3944700 3944701 3944701
97044-328EA 2306.8 CAD
97044-328 97044-330 470203-202
Guardian™ 500 Airflow Monitor with Epoxy-Coated Steel Casing, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
The Guardian 500 Airflow Monitor is designed to continuously monitor airflow through Labconco fume hoods and other enclosures
Compatible with the following units: Protector XVS Ventilation Stations, Protector Demonstration Hoods, Protector Work Stations without Blower
This permanently-installed device provides both visual and audible alarms to alert the user of abnormal airflow conditions. From the monitors face plate, the user can easily select and calibrate a setpoint between 30 and 250 fpm using a velocity meter and screwdriver. A green Safe light indicates normal flow conditions. When flow conditions lower than the set point are encountered, a red Low is activated and an audible alarm sounds. If configured, an amber Sash High light flashes when the sash is raised above the safe working opening. Pressing the Mute button temporarily silences the alarm. The monitor has a built-in sensor, three relay inputs and one relay output. The relay inputs can be configured for night setback, external alarm and sash high alarm. Guardian 500 Airflow Monitor Kits for use with Protector XVS Ventilation Stations, Protector Demonstration Hoods and Protector Work Stations include the air monitor, electrical power pack, tubing, airflow tube adapter and glacier white epoxy-coated steel casing.