Hooded Combination Kjeldahl Digestion/Distillation Units, Labconco
Supplier: Labconco
2117613 2117712 2117713 2117812 2117813 2117611 2117612
10129-200EA 107171.4 CAD
10129-200 10129-202 10129-204 10129-206 10129-208 10129-262 10129-264
Hooded Combination Kjeldahl Digestion/Distillation Units, Labconco
Digestion Systems Mineralisation Workstations
Hooded Combination Kjeldahl Digestion/Distillation Apparatus
Combines macro digestion and distillation processes for nitrogen determination of samples in one space-saving unit. Hood surrounding the apparatus helps contain the processing heat and vent it from the laboratory. Able to digest and distill up to six, twelve, or eighteen samples at one time. Hood is constructed of epoxy-coated formed steel with horizontal-sliding, tempered safety glass sashes. Frame is constructed of powder-coated tubular steel. Remote blowers are not included.
Ordering information: Requires remote exhaustMore Product Information