Sodium chloride 0,9% water Normal saline solution

Supplier: G-Biosciences

Synonyms: Saline, NaCl, Sea salt, Halite

786-560 786-561
CA89167-772EA 104.98 CAD
CA89167-772 CA89167-774
Sodium chloride 0,9% water Normal saline solution
Sodium chloride

G-Biosciences offers 0.9% NaCl in two convenient sizes to meet your particular laboratory needs. These sterile saline solutions are suitable for numerous applications.

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR