Urea, Ultra Pure Grade

Supplier: G-Biosciences

Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide

BC89 RC-112
CA82022-850EA 342.92 CAD
CA82022-850 CA71003-500
Urea, Ultra Pure Grade
Urea
Formula: NH₂CONH₂
MW: 60.06 g/mol
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 132.7 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR