Tricine

Supplier: G-Biosciences

Synonyms: 2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine

RC-103 RC-104
CA71003-486EA 256.23 CAD
CA71003-486 CA71003-488
Tricine
Tricine

G-Biosciences' tricine is a buffer component used for the separation of low molecular weight peptides.

Formula: (HOCH₂)₃CNHCH₂CO₂H
MW: 179.17 g/mol
Melting Pt: 187 °C
Density: 1.523 g/cm³ (19.85 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004277
CAS Number: 5704-04-1
EINECS: 227-193-6
Merck Index: 13,09724

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR