Sodium dihydrogen phosphate
Supplier: G-Biosciences
RC-097
RC-098
CA71003-476EA
158.19
CAD
CA71003-476
CA71003-478
Sodium dihydrogen phosphate
Sodium dihydrogen phosphate
Formula:
NaH₂PO₄ Melting Pt: ∼650 °C Density: 0.9996 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003527 CAS Number: 7558-80-7 EINECS: 231-449-2 REACH: 01-2119489796-13 Merck Index: 12,08806 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...