Sodium chloride for biochemistry

Supplier: G-Biosciences

Synonyms: Saline, NaCl, Sea salt, Halite

RC-093 RC-094
CA95029-574EA 190.6 CAD
CA95029-574 CA71003-470
Sodium chloride for biochemistry
Sodium chloride
Formula: NaCl
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR