Potassium dihydrogen phosphate

Supplier: G-Biosciences

RC-083 RC-084
CA71003-454EA 276.91 CAD
CA71003-454 CA71003-456
Potassium dihydrogen phosphate
Potassium dihydrogen phosphate
Formula: KH₂PO₄
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: 253 °C
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR