Potassium chloride
Supplier: G-Biosciences
RC-123
RC-124
CA71003-520EA
245.66
CAD
CA71003-520
CA71003-522
Potassium chloride
Potassium chloride
Formula:
KCl Boiling Pt: 1420 °C (1013 hPa) Melting Pt: 773 °C Density: 1.98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011360 CAS Number: 7447-40-7 EINECS: 231-211-8 Merck Index: 14,07621 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...