Polyethylene glycol 8,000

Supplier: G-Biosciences

Synonyms: PEG 8,000, Ethylene glycol polymer 8000, PEG 8000

RC-077 RC-078
CA71003-442EA 193.33 CAD
CA71003-442 CA71003-444
Polyethylene glycol 8,000
Polyethylene glycol 8,000
Formula: H(OCH₂CH₂)nOH
MW: 8000 g/mol
Boiling Pt: 250
Density: 1.125 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD01779615
CAS Number: 25322-68-3
EINECS: 500-038-2
Merck Index: 12,07729

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR