γ-(N-Morpholino)propanesulfonic acid (MOPS)
Supplier: G-Biosciences
Synonyms:
3-Morpholinopropane-1-sulfonic acid, 3-(N-Morpholino)propanesulfonic acid, 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
RC-072
RC-073
CA71003-438EA
277.32
CAD
CA71003-438
CA71003-440
γ-(N-Morpholino)propanesulfonic acid (MOPS)
γ-(N-Morpholino)propanesulphonic acid (MOPS)
Formula:
C₇H₁₅NO₄S MW: 209.27 g/mol Melting Pt: > 250 °C Flash Pt: 110 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00006183 CAS Number: 1132-61-2 EINECS: 214-478-5 Merck Index: 13,06288 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...