D-Methionine for biochemistry
Supplier: G-Biosciences
Synonyms:
D(+)-Methionine, (R)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-2-Amino-4-(methylthio)butyric acid, (R)-(+)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-Methionine, H-D-Met-OH
RC-070
RC-071
CA71003-434EA
151
CAD
CA71003-434
CA71003-436
D-Methionine for biochemistry
D-Methionine
Formula:
C₅H₁₁NO₂S Boiling Pt: 278 °C (1013 hPa) Melting Pt: 136 °C |
MDL Number:
MFCD00002622 CAS Number: 348-67-4 EINECS: 206-483-6 Merck Index: 13,06004 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...