Magnesium chloride hexahydrate for biochemistry

Supplier: G-Biosciences

Synonyms: Magnesium dichloride

RC-068 RC-069
CA71003-430EA 151 CAD
CA71003-430 CA71003-432
Magnesium chloride hexahydrate for biochemistry
Magnesium chloride hexahydrate
Formula: MgCl₂·6H₂O
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 117 °C
Density: 1.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR