L(+)-Lysine monohydrochloride
Supplier: G-Biosciences
Synonyms:
(S)-(+)-2,6-Diaminohexanoic acid monohydrochloride
, H-Lys-OH.HCl
, L-Lysine monohydrochloride
, H-Lys-OH·HCl
, L-Lysine monohydrocholoride
, (S)-(+)-Lysine monohydrochloride
, Lysine Hydrochloride
, L-Lysine Hydrochloride
, (S)-(+)-2,6-Diaminocaproic acid monohydrochloride
RC-066
RC-067
RC-066
RC-067
CA71003-426EA
140.45
CAD
71003-426
71003-428
CA71003-426
CA71003-428
L(+)-Lysine monohydrochloride
L(+)-Lysine monohydrochloride
Formula:
H₂N(CH₂)₄CH(NH₂)CO₂H·HCl MW: 182.65 g/mol |
MDL Number:
MFCD00064564 CAS Number: 657-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...