HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid free acid
Supplier: G-Biosciences
RC-060
RC-061
RC-121
CA71003-416EA
165.27
CAD
CA71003-416
CA71003-418
CA71003-516
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid free acid
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)
Formula:
C₈H₁₈N₂O₄S Melting Pt: 210…215 °C Density: 1.448 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006158 CAS Number: 7365-45-9 EINECS: 230-907-9 Merck Index: 13,04672 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...