Guanidinium thiocyanate

Supplier: G-Biosciences

Synonyms: Guanidinium sulfocyanideGuanidine ThiocyanateGuanidinium sulphocyanideGTCGuanidinium rhodanide

RC-059 RC-058 RC-059 RC-058
CA71003-414EA 1052.79 CAD
CA71003-414 71003-412 CA71003-412 71003-414
Guanidinium thiocyanate
Guanidinium thiocyanate
Formula: CH₅N₃·CHNS
MW: 118.16 g/mol
Melting Pt: 118 °C
Density: 1.29 g/cm³ (20 °C)
MDL Number: MFCD00013027
CAS Number: 593-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR