Guanidinium hydrochloride

Supplier: G-Biosciences
Warning

BC88 BC85 RC-057
CA82022-848EA 950.88 CAD
CA82022-848 CA71003-304 CA71003-410
Guanidinium hydrochloride
Guanidine hydrochloride

A selection of strong chemicals to aid in the denaturation of proteins. Denaturants, or chaotropes, disrupt water interactions resulting in the solubilization of hydrophobic proteins and peptides. The chaotropes also act on all proteins to unfold them and alter their three-dimensional structure.


Available as a powder or 8M solution

Formula: NH₂C(=NH)NH₂·HCl
Melting Pt: 185 °C
Density: 1.354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR