Diethyl pyrocarbonate
Supplier: G-Biosciences
|
Warning
|
RC-041
RC-042
CA71003-388EA
172.01
CAD
CA71003-388
CA71003-390
Diethyl pyrocarbonate
Diethyl pyrocarbonate
Formula:
O(COOC₂H₅)₂ Boiling Pt: 93 °C (1013 hPa) Density: 1.123 g/cm³ (20 °C) Flash Pt: 69 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009106 CAS Number: 1609-47-8 EINECS: 216-542-8 Merck Index: 13,08088 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...