L-Alanine

Supplier: G-Biosciences

Synonyms: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine

RC-013 RC-014
CA71003-340EA 277.32 CAD
CA71003-340 CA71003-342
L-Alanine
L-Alanine
Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 189
Melting Pt: 79
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064410
CAS Number: 56-41-7
EINECS: 200-273-8
Merck Index: 13,00203

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR