Acrylamide, powder for biochemistry

Supplier: G-Biosciences
Danger

Synonyms: Prop-2-enamide

RC-001 RC-002
CA95029-560EA 350.24 CAD
CA95029-560 CA71003-326
Acrylamide, powder for biochemistry
Acrylamide (monomer)
Formula: C₃H₅NO
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 84.5 °C
Density: 1.122 g/cm³ (20 °C)
Flash Pt: 138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008032
CAS Number: 79-06-1
EINECS: 201-173-7
UN: 2074
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7687b7f17a8cdf31c559a33fd5d4021a2e50a40a&vid=fecebe0c34b037788df119c11bfc9637d78cda0d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR