West Condenser, No Hold-Up, Jacketed, Ace-Threds, Ace Glass Incorporated
Supplier: Ace Glass
6029-112 6029-115 6029-117 6029-118 6029-119
89054-004EA 457.13 CAD
89054-004 89054-006 89054-008 89054-010 89054-012
West Condenser, No Hold-Up, Jacketed, Ace-Threds, Ace Glass Incorporated
Condensers
Jacketed West Condenser with standard taper 24/40 inner and reinforced outer joints at bottom and top
Has 7 Ace-Safe hose connections for 1/4" I.D. tubing.
Ordering information: Ace-Thred connectors included. Version with hose barbs rather than Ace-Threds is available. Contact your Avantor representative for further information.