Glutathione, Reduced

Supplier: G-Biosciences

786-587 786-588
CA71003-206EA 160.8 CAD
CA71003-206 CA71003-208
Glutathione, Reduced
Enzymes

G-Biosciences offers reduced glutathione (CAS # 70-18-8), otherwise known as GSH and γ-L-Glutamyl-L-cysteinyl-glycine.


Reduced glutathione has a molecular weight of 307.32, and it is freely soluble in water (50 mg per mL), dilute alcohol, liquid ammonia, and DMF.


The amino acid sequence of reduced glutathione is γ-Glu-Cys-Gly. Its linear formula is C₁₀H₁₇N₃O₆S.


Ordering information: Accessories available.

Order Now

SPECIFICATIONS

Enzyme name Glutathione, Reduced
Environmentally Preferable
Source
Species

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR