Pearl™ IgG Purification, G-Biosciences
Supplier: G-Biosciences
786-798 786-799 786-800 786-801
CA89168-052EA 1232.49 CAD
CA89168-052 CA89168-054 CA89168-056 CA89168-058
Pearl™ IgG Purification, G-Biosciences
Protein Purification Protein Purification Resins
G-Biosciences' Pearl™ IgG Purification Resin is designed for the one-step purifiication of the immunoglobulin G (IgG) antibodies from serum
G-Biosciences' Pearl™ IgG Purification Resin purifies IgG in less than 15 minutes, which is more rapid than the commonly used Protein A and Protein G resins. The performance of the Pearl™ IgG Purification Resin is comparable or better than the Protein A and Protein G resins, with a high recovery of over 90% and purity greater than 80%.
G-Biosciences' Pearl™ IgG Purification Resin is also available as a complete kit, and as a spin format kit. The Pearl™ IgG Purification (Spin Format) kit is ideal for the rapid, small scale purification of IgG. The kit is supplied with 3 mL Pearl™ IgG Purification Resin, IgG Isolation Buffer and 20 spin columns. It is suitable for purifying up to 25 mg IgG. The kit is supplied with 25 mL Pearl™ IgG Purification Resin and IgG Isolation Buffer and is suitable for the isolation of IgG from ~100 mL serum (~200 mg IgG).
Pearl™ IgG Purification Resin binds the high abundant, non-IgG proteins (i.e albumin) and allows the IgG molecules to pass through in a physiological buffer. The purified IgG molecules can be stored or used in further downstream applications without further clean-up, such as ammonium sulfate precipitation.
Ordering information: Accessories included. Additional supplies available.