ART® Barrier Ultra Micro Tips with MicroPoint Design, Thermo Scientific
Supplier: Thermo Scientific
2140 2065-HR 2140-HR 2065E-HR 2065E-05-HR 2179-HR 2179-05-HR 2140-RI
53509-500CS 1880.79 CAD
53509-500 89082-068 89082-070 89082-078 89082-080 89082-090 89082-092 89082-106
ART® Barrier Ultra Micro Tips with MicroPoint Design, Thermo Scientific
Pipette Tips
Molecular Bioproducts ART® pipette tips are pre-sterilized and precision molded with the ART® self-sealing barrier.
They are guaranteed to prevent carryover contamination during aspiration and delivery of samples. Ideally suited for low volume applications in genetic studies, forensics, PCR, and radioisotope sampling.