HOOK™ Activated Agarose (Amine Reactive) for Affinity Column Generation, G-Biosciences
Supplier: G-Biosciences
786-063 786-066
CA82022-398EA 1929.16 CAD
CA82022-398 CA82022-404
HOOK™ Activated Agarose (Amine Reactive) for Affinity Column Generation, G-Biosciences
Protein Linking and Labeling Systems
G-Biosciences' amine reactive HOOK™ Activated Agarose is 6% agarose that has been activated to generate reactive aldehyde groups
G-Biosciences' amine reactive HOOK™ Activated Agarose is designed for the generation of peptide and protein affinity columns, for the purification of antibodies and for the discovery of important interacting proteins and cofactors. G-Biosciences offers activated agarose for the coupling of peptides and proteins to agarose through their primary amines.
G-Biosciences' amine reactive HOOK™ Activated agarose is also supplied in a complete kit for the generation of 5 x 2 mL resins. The kit is supplied with all the necessary reagents and columns. Binding capacity: 20 mg protein per mL resin.
The aldehyde groups of the agarose react spontaneously with primary amines, located at the N-terminus of proteins or in lysine residues, to form intermediate Schiff Base complexes. These, in turn, are selectively reduced by reductive amination to form stable amine linkages between the agarose and the ligand.
Ordering information: Accessories included. Additional supplies available.