Tris-Glycine, G-Biosciences
Supplier: G-Biosciences
786-477 786-478
CA95029-258EA 186.63 CAD
CA95029-258 CA95029-260
Tris-Glycine, G-Biosciences
Buffers Electrophoresis Buffers Running Buffers
G-Biosciences' Tris-Glycine [10X] contains: 0.25M Tris and 1.92M Glycine, at a pH of 8.3.
Ordering information: Accessories available.