Gram Negative Lysis Buffer, G-Biosciences
Supplier: G-Biosciences
786-579 786-580 786-566 786-567
CA71003-190EA 572.13 CAD
CA71003-190 CA71003-192 CA89167-784 CA89167-786
Gram Negative Lysis Buffer, G-Biosciences
Buffers Extraction Buffers
G-Biosciences' Gram-negative Lysis Buffer is an extraction buffer for soluble proteins from Gram-negative bacteria
Depending on the application, additional agents such as reducing agents, chelating agent, nuclease and protease inhibitors cocktail may be added into Gram-negative lysis buffer. This buffer has been tested for use with several widely used bacteria including E. coli strains.
It is a proprietary improvement on the lysozyme based lysis in combination with various salts and agents, which allows extraction of soluble proteins.
Ordering information: Accessories available.
Order Now
SPECIFICATIONS
Environmentally Preferable