Destain solutions, G-Biosciences
Supplier: G-Biosciences
786-526
786-527
CA95043-514EA
241.83
CAD
CA95043-514
CA95043-516
Destain solutions, G-Biosciences
Electrophoresis Reagents
Stain Removal Reagents
G-Biosciences' Destain I contains approximately 45% methanol and 9% glacial acetic acid. G-Biosciences' Destain II contains approximately 5% methanol and 7% acetic acid.