Nalgene® Closures for Dropper Bottles, Thermo Scientific
Supplier: Thermo Fisher Scientific
312760-0000 312760-0010 312760-0020 312760-0040 312760-0050 312760-0060
46300-568CS 622.55 CAD
46300-568 46300-570 46300-572 46300-574 46300-576 73521-300
Nalgene® Closures for Dropper Bottles, Thermo Scientific
Bottles Dropping Bottles
Nalgene closures for dropper bottles fit 312750 and 312751 bottles and must be ordered with dispensing tip to complete Dropper Bottle System.
The closures are available in natural, white, yellow, green, red, and blue. Neck finish for 46300-568 and 46300-570 is 15-415.