Water endotoxin-free

Supplier: G-Biosciences

Synonyms: Hydrogen oxide

786-670 786-671
CA89167-874EA 91.21 CAD
CA89167-874 CA89167-876
Water endotoxin-free
Water

G-Biosciences' Endotoxin-Free Water is free of all endotoxins and enzymes, including proteases. Certified tested by the Limulus amebocyte lysate (LAL) test for endotoxins and determined to be <0.0050 EU per milliliter.


G-Biosciences also offers Molecular-Grade, DEPC-Treated, and Proteomic-Grade Water. Accessories Available.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR