Water, molecular grade DNAse_RNAse_free

Supplier: G-Biosciences

Synonyms: Hydrogen oxide

786-292 786-293
CA95043-414EA 131.76 CAD
CA95043-414 CA95043-416
Water, molecular grade DNAse_RNAse_free
Water

G-Biosciences' Molecular Grade Water™ is suitable for use in molecular biology applications, PCR reactions, 2D electrophoresis and mass spectrometry.


G-Biosciences' Molecular Grade Water™ is suitable for use in molecular biology applications that demand a high quality of water and assurance that the water is free from DNase, RNase and protease contamination. No toxic agents, such as DEPC, are used in the manufacturing of Molecular Grade Water™, eliminating DEPC interference of enzymatic reactions. Each lot is quality tested for the absence of RNase, DNase and protease contamination.


G-Biosciences also offers Endotoxin-Free, DEPC-Treated, and Proteomic-Grade Water.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR