Finntip® Flex Filter Tips, Thermo Scientific
Supplier: Thermo Fisher Scientific
94056510 94056520 94056580 94056810 94056970 94056380 94056980 94056710
89047-106CS 466.35 CAD
89047-106 89047-108 89047-110 89047-114 89049-826 89087-480 89087-482 89047-112
Finntip® Flex Filter Tips, Thermo Scientific
Pipette Tips
The Finntip Flex Filter Pipette Tips combine the ergonomics of a standard Finntip Flex pipette tip and the protection provided by a filtered tip.
Finntip flex filter pipette tips effectively minimize the risk of passing contamination from the pipette to the sample. Including a range of products certified free from DNA, DNase, RNase and endotoxin, these sterile tips suit all sensitive procedures, including PCR-based reactions where contamination is a concern. The filters are composed of an inert non-sealing polyethylene matrix and are effective at eliminating carryover contamination.
Order Now
SPECIFICATIONS
Environmentally Preferable