Anti-Avidin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
200-4397-0100 200-4397-0100
CA20043970100EA 754.01 CAD
CA20043970100 RL200-4397-01
Anti-Avidin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Avidin Antibody Peroxidase Conjugated has been assayed against 1.0 ug of Avidin [Hen Egg White ] in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temp

  • Recommended Dilutions: ELISA: 1:10000 - 1:50000; Immunofluorescence: 1:500 - 1:2000; Western Blot: 1:2000 - 1:5000

Type: Tag
Antigen: Avidin
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR