Anti-Horse Serum Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
108-4101 108-4101
CA108-4101EA 512 CAD
RL108-4101 CA108-4101
Anti-Horse Serum Rabbit Polyclonal Antibody
Antibodies
Anti-Horse serum antibody is suitable for western blotting, IP and for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Horse Serum
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Horse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR