Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Rockland Immunochemicals
609-1503 609-1503
CA609-1503EA 406 CAD
RL609-1503 CA609-1503
Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Secondary Goat Anti-IgG F(c) Reacts with Human

Recommended Dilutions: ELISA: 1:5000 - 1:10000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR