Anti-MTOR Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-422
CA600-401-422EA 834 CAD
CA600-401-422
Anti-MTOR Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in immunochemistry, ELISA and western blotting. Western blotting shows reactivity specific for phospho mTOR detecting a band at approximately 250 kDa.

Recommended Dilutions: ELISA: 1:10000 - 1:100000; Western Blot: 1:500 - 1:2000; Immunochemistry: 5.0 µg/ml;

Type: Primary
Antigen: MTOR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR